CELLTREAT Scientific Product Sterile Cell Strainers
Designed to help isolate cells and remove debris and clumps
  • Pair directly with your 50-mL centrifuge tubes
  • Color-coded housing helps differentiate sizes
  • Extended gripping tab makes handling easy
Polypropylene housing with nylon mesh filter eliminates the need to create filters from gauze, paper, or other materials. Elongated lip on housing enable aseptic handling with forceps. Strainers are supplied sterile (gamma irradiated). Choose from 40, 70, or 100 µm mesh pore size strainers.
34,700.00 - ₹36,200.00INR / Case of 50
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (micron) Description
Availability Pricing
CELLTREAT Scientific Products 229481 Sterile Cell Strainers, 40 µm, Blue, Individually Wrapped; 50/cs 0633651 Mfr # 229481
COMPARE
40 Sterile Cell Strainers, 40 µm, Blue, Individually Wrapped; 50/cs
₹36,200.00INR / Case of 50
CELLTREAT Scientific Products 229482 Sterile Cell Strainers, 40 µm, Blue, Bulk Packed; 50/cs 0633653 Mfr # 229482
COMPARE
40 Sterile Cell Strainers, 40 µm, Blue, Bulk Packed; 50/cs
₹34,700.00INR / Case of 50
CELLTREAT Scientific Products 229483 Sterile Cell Strainers, 70 µm, White, Individually Wrapped, 50/cs 0633655 Mfr # 229483
COMPARE
70 Sterile Cell Strainers, 70 µm, White, Individually Wrapped, 50/cs
₹35,800.00INR / Case of 50
CELLTREAT Scientific Products 229484 Sterile Cell Strainers, 70 µm, White, Bulk Packed; 50/cs 0633657 Mfr # 229484
COMPARE
70 Sterile Cell Strainers, 70 µm, White, Bulk Packed; 50/cs
₹34,700.00INR / Case of 50
CELLTREAT Scientific Products 229485 Sterile Cell Strainers, 100 µm, Yellow, Individually Wrapped; 50/cs 0633659 Mfr # 229485
COMPARE
100 Sterile Cell Strainers, 100 µm, Yellow, Individually Wrapped; 50/cs
₹36,200.00INR / Case of 50
CELLTREAT Scientific Products 229486 Sterile Cell Strainers, 100 µm, Yellow, Bulk Packed; 50/cs 0633661 Mfr # 229486
COMPARE
100 Sterile Cell Strainers, 100 µm, Yellow, Bulk Packed; 50/cs
₹35,000.00INR / Case of 50