Cole-Parmer 316 Stainless Steel three-blade impeller, 2" dia x 3/8" bore dia
Representative image only.
Cole-Parmer 316 Stainless Steel three-blade impeller, 2" dia x 3/8" bore dia
Cole-Parmer | Item: 0455255
Keep your overhead mixer running smoothly
₹13,900.00INR / Each
₹13,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer 316 Stainless Steel three-blade impeller, 2" dia x 3/8" bore dia
Cole-Parmer 316 Stainless Steel three-blade impeller, 2" dia x 3/8" bore dia
0455255
₹13,900.00INR / Each
+ + + +
Caframo High-Torque Brushless Mixer; 12 to 1800 rpm, 115 VAC
5080000
₹476,000.00INR / Each
+
Cole-Parmer   GP Mixer Set; 120/230 VAC
5500800
₹251,000.00INR / Each
+
Cole-Parmer Heavy-duty clamp holds rods up to 1-1/8" dia
0456124
₹16,400.00INR / Each
+
Cole-Parmer Replacement Keyless Chuck for SP and SD Mixers
5500810
₹42,600.00INR / Each
+
Caframo Straight Blade Impeller, SS, 32 mm (1-1/4") Dia
5080151
₹44,400.00INR / Each
+
Cole-Parmer Heavy-Duty Stand for Overhead Mixer
9996821
₹202,000.00INR / Each
=
Total Price 1309200.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Update clamps, mounts, shafts and impellers to accommodate new protocols. These replacement parts will keep your overhead mixer running smoothly.
Three-blade impellers are for general-purpose mixing. Use alone or mount several on a single shaft for thorough agitation.
Two year warranty
Specifications
  • Description316 Stainless Steel three-blade propeller, 2" dia x 3/8" bore dia