Cole-Parmer Heavy-duty clamp holds rods up to 1-1/8" dia
Representative image only.
Cole-Parmer Heavy-duty clamp holds rods up to 1-1/8" dia
Cole-Parmer | Item: 0456124
Enhance the performance of your Cole-Parmer mixer
₹16,400.00INR / Each
₹16,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Heavy-duty clamp holds rods up to 1-1/8" dia
Cole-Parmer Heavy-duty clamp holds rods up to 1-1/8" dia
0456124
₹16,400.00INR / Each
+
Cole-Parmer 316 Stainless Steel Shaft, 5/16" dia x 18" L
0455230
₹19,300.00INR / Each
+
Caframo High-Torque Brushless Mixer; 12 to 1800 rpm, 115 VAC
5080000
₹476,000.00INR / Each
+
Caframo Straight Blade Impeller, SS, 32 mm (1-1/4") Dia
5080151
₹44,400.00INR / Each
+
Caframo Sweep Blade, Silicone-Molded; 6/Pk
5080170
₹21,800.00INR / Pkg of 1
=
Total Price 577900.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Compatible with Cole-Parmer Compact Digital Lab Mixer (50801-00)
  • Chemical-resistant epoxy construction for harsh applications
  • 2 year warranty
More About this Item
Heavy-duty clamp, made of cast aluminum zinc alloy with chemical-resistant epoxy coating, holds rods up to 1-1/8" dia. Handles made of Stainless Steel.
Two year warranty
Specifications
  • DescriptionHeavy-duty clamp holds rods up to 1-1/8" dia