Cole-Parmer 300 pH/ORP/Temperature 1/8-DIN Controller

Representative image only.

Cole-Parmer 300 pH/ORP/Temperature 1/8-DIN Controller

Cole-Parmer | Item: 0580212

Automatic buffer recognition for quicker setup

₹45,800.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹45,800.00INR / Each

Buy It Together

Current Item

Cole-Parmer 300 pH/ORP/Temperature 1/8-DIN Controller
Cole-Parmer 300 pH/ORP/Temperature 1/8-DIN Controller
0580212

₹45,800.00INR / Each
+ + +
Cole-Parmer Epoxy pH Electrode BNC
0599223

₹23,200.00INR / Each
+ + + + + =
Total Price 482300.00 ADD 9 ITEMS TO CART
Product Description

Key Features

  • Backlight LCD for clear measurement display
  • Automatic buffer recognition of US and NIST buffers
  • Universal power supply input from 100 to 240VAC
  • Easy-to-install 1/8-DIN compact plastic casing
  • Two SPDT relays offer high/low control to activate an alarm, a pump, or another device
  • 1 year warranty

More About this Item

Cole-Parmer pH/ORP controllers offer features found in more expensive models. Automatic buffer recognition, absolute and relative ORP measurements, and automatic temperature compensation ensure measurement accuracy. Models with two sets of programmable on/off relays are ideal for high/low control to activate an alarm, a pump, or another device. Models with an isolated 4-20 mA output signal are suitable for opening valves and pacing pumps. ATC element is user selectable, 10K NTC or 1000 Ohm RTD. Order 56105-60 BNC-to-Stripped Wire Adapter for easy connections on terminal strip on back of controller.

1 Year Warranty
Specifications
  • Control TypeOn/Off
  • Display TypeLCD
  • Housing Material1/8-DIN
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01
  • pH Accuracy±2% of reading
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1mV
  • mV Accuracy±2mV
  • Min Temperature (° C)-10
  • Max Temperature (° C)120
  • Min Temperature (° F)14
  • Max Temperature (° F)248
  • Temperature Resolution0.1°
  • Temperature Accuracy±0.5°C
  • Temperature CompensationAutomatic
  • Output Relay2 SPDT relays
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Width (cm)9.6
  • Height (cm)4.8
  • Depth (cm)11
  • Width (in)3 21/64
  • Height (in)1 3/4
  • Depth (in)4 1/4
  • Panel Mount Size1/8-DIN
  • Description300 pH/ORP/Temperature 1/8-DIN Controller; with Two SPDT relays
  • Warranty1 year
Reviews
Q&A