Cole-Parmer BNC-to-Stripped Wire Adapter
Representative image only.
Cole-Parmer BNC-to-Stripped Wire Adapter
Cole-Parmer | Item: 5610560
Get the most out of water analysis equipment
₹3,600.00INR / Each
₹3,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer BNC-to-Stripped Wire Adapter
Cole-Parmer BNC-to-Stripped Wire Adapter
5610560
₹3,600.00INR / Each
+
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Transmitter
0580214
₹46,300.00INR / Each
+ + + =
Total Price 213800.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Replace or upgrade electrodes, probes, cells, and other accessories
More About this Item
Water analysis accessories for Cole-Parmer® meters. Many of these items are compatible with other brands of meter.
Use this female BNC-to-stripped wire end connector to simplfy connection and replacement of pH/ORP electrodes with male BNC connections to transmitters and controllers with terminal block connectors.
Specifications
  • DescriptionBNC-to-Stripped Wire Adapter