Oakton self-cleaning pH electrode, CPVC housing, 10-ft cable, BNC
Representative image only.
Oakton self-cleaning pH electrode, CPVC housing, 10-ft cable, BNC
No more solid buildups
₹26,300.00INR / Each
₹26,300.00INR / Each
Buy It Together
Current Item
Oakton self-cleaning pH electrode, CPVC housing, 10-ft cable, BNC
Oakton self-cleaning pH electrode, CPVC housing, 10-ft cable, BNC
2700300
₹26,300.00INR / Each
+
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Transmitter
0580214
₹46,300.00INR / Each
+ +
Cole-Parmer Epoxy pH Electrode BNC
0599223
₹23,200.00INR / Each
+ + + + =
Total Price 346900.00 ADD 8 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Flat surface prevents buildups, while fluid flow cleans surface for longer life
  • Use with most meters and controllers—BNC connector and ATC element work with a variety of units
  • Double junction for difficult applications
  • 1 year warranty
More About this Item
Choose either CPVC housing or chemical-resistant Ryton® PPS housing
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialCPVC
  • Min pH (pH)12
  • Max Temperature (° C)80
  • Max Pressure (PSI)100
  • Thread Size NPT(M) (in)3/4
  • ATC ElementNone
  • ConnectorBNC
  • Cable Length (m)3
  • Descriptionself-cleaning pH electrode, CPVC housing, 10-ft cable, BNC