Cole-Parmer 1" NPT(F) T-fitting with 3/4" NPT(F), CPVC adapter installed in branch
Representative image only.
Cole-Parmer 1" NPT(F) T-fitting with 3/4" NPT(F), CPVC adapter installed in branch
Cole-Parmer | Item: 2700380
Replace cells economically
₹16,300.00INR / Each
₹16,300.00INR / Each
Product Description
Key Features
  • In-line Fittings
  • 1" NPT(F) T-fittings with adapter
  • Self-cleaning electrode in 27003-series — 5 to 80ºC
  • 1 year warranty
More About this Item
In-line fittings make switching out electrodes simple.
This is a 1" NPT(F) connection on all three sides and the top has a 3/4" NPT(F) for fitting - reducing bushing installed.
1 Year Warranty
Specifications
  • MaterialCPVC
  • Description1" NPT(F) T-fitting with 3/4" NPT(F), CPVC adapter installed in branch