BYK-Gardner PV-0174 Ford Viscosity Cup #4, 70 to 370 Centistokes
Representative image only.
BYK-Gardner PV-0174 Ford Viscosity Cup #4, 70 to 370 Centistokes
BYK-Gardner | Mfr: PV-0176| Item: 0871120
Use accessory stand with bath ports to measure viscosity at various or critcal temperatures
₹74,400.00INR / Each
₹74,400.00INR / Each
Buy It Together
Current Item
BYK-Gardner PV-0174 Ford Viscosity Cup #4, 70 to 370 Centistokes
BYK-Gardner PV-0174 Ford Viscosity Cup #4, 70 to 370 Centistokes
0871120
₹74,400.00INR / Each
+
Gardco EZ1 Series Viscosity Cup #1, 10 to 36 Centistokes
0870000
₹60,800.00INR / Each
+
Cole-Parmer Viscosity Cup #2, 40 to 230 centistokes
0870061
₹25,500.00INR / Each
+ +
BYK-Gardner PV-7208 Ford Viscosity Cup Accessory Kit
0871150
₹36,600.00INR / Each
=
Total Price 264800.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • ±2% production tolerance
  • Efflux time is 20 - 105 seconds
  • Orifice diameter of 0.162"
  • 1 year warranty
More About this Item
These new Ford viscosity cups are extremely easy to use. Simply pour your sample into the cup and measure the time it takes for the liquid to flow through the orifice until the first break in the liquid stream. Use the included table to convert the elapsed time to centistokes (cSt).
The cups are made from solid aluminum; the orifice is made from brass. All the cups are calibrated to NIST-traceable oils.
An accessory kit, a stand, and a stand with water bath ports are sold separately.
1 Year Warranty
Specifications
  • Sample Volume Needed (mL)135
  • Cup Number4
  • Nist-TraceableNo
  • DescriptionFord Viscosity Cup #4, 70 to 370 Centistokes