BYK-Gardner PV-7208 Ford Viscosity Cup Accessory Kit
Representative image only.
BYK-Gardner PV-7208 Ford Viscosity Cup Accessory Kit
BYK-Gardner | Mfr: PV-7208| Item: 0871150
Optimize performance of your BYK-Gardner ford viscosity cup
₹36,600.00INR / Each
₹36,600.00INR / Each
Buy It Together
Current Item
BYK-Gardner PV-7208 Ford Viscosity Cup Accessory Kit
BYK-Gardner PV-7208 Ford Viscosity Cup Accessory Kit
0871150
₹36,600.00INR / Each
+ =
Total Price 111000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Keep your ford viscosity cup running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your BYK-Gardner ford viscosity cups.
Ford cup accessory kit includes cover glass for removing excess sample from the cup, bubble level for leveling cup and stand, stainless steel beaker, and package of cleaning swabs.
1 Year Warranty
Specifications
  • DescriptionFord Viscosity Cup Accessory Kit