Ford Viscosity Cups
Use accessory stand with bath ports to measure viscosity at various or critcal temperatures
  • ±2% production tolerance
These new Ford viscosity cups are extremely easy to use. Simply pour your sample into the cup and measure the time it takes for the liquid to flow through the orifice until the first break in the liquid stream. Use the included table to convert the elapsed time to centistokes (cSt).
The cups are made from solid aluminum; the orifice is made from brass. All the cups are calibrated to NIST-traceable oils.
An accessory kit, a stand, and a stand with water bath ports are sold separately.
74,400.00 - ₹137,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Cup Number Nist-Traceable
Availability Pricing
BYK-Gardner PV-7201 Ford Viscosity Cup #2, 25 to 120 Centistokes 0871100 Mfr # PV-7201
COMPARE
2 No
₹74,400.00INR / Each
BYK-Gardner SC-7201 Ford Viscosity Cup #2 with Certificate, 25 to 120 Centistokes 0871105 Mfr # PV-0172
COMPARE
2 Yes
₹137,000.00INR / Each
BYK-Gardner PV-0173 Ford Viscosity Cup #3, 37 to 231 Centistokes 0871110 Mfr # PV-0175
COMPARE
3 No
₹74,400.00INR / Each
BYK-Gardner SC-0173 Ford Viscosity Cup #3 with Certificate, 37 to 231 Centistokes 0871115 Mfr # PV-0173
COMPARE
3 Yes
₹137,000.00INR / Each
BYK-Gardner PV-0174 Ford Viscosity Cup #4, 70 to 370 Centistokes 0871120 Mfr # PV-0176
COMPARE
4 No
₹74,400.00INR / Each
BYK-Gardner SC-0174 Ford Viscosity Cup #4 with Certificate, 70 to 370 Centistokes 0871125 Mfr # PV-0174
COMPARE
4 Yes
₹137,000.00INR / Each