BYK-Gardner PV-8202 Zahn Viscosity Cup, number 2, 30 to 230 cSt, 22 to 80 sec
Representative image only.
BYK-Gardner PV-8202 Zahn Viscosity Cup, number 2, 30 to 230 cSt, 22 to 80 sec
BYK-Gardner | Mfr: PV-8202| Item: 0871132
₹67,500.00INR / Each
₹67,500.00INR / Each
Buy It Together
Current Item
BYK-Gardner PV-8202 Zahn Viscosity Cup, number 2, 30 to 230 cSt, 22 to 80 sec
BYK-Gardner PV-8202 Zahn Viscosity Cup, number 2, 30 to 230 cSt, 22 to 80 sec
0871132
₹67,500.00INR / Each
+
Gardco VI-3403 Standard Ford Dip Viscosity Cup, orifice #3
0870900
₹142,000.00INR / Each
+ =
Total Price 283900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Stainless steel cup with precsion-drilled orifice for quick, easy use in shops, factories, and laboratories.
Specifications
  • Viscosity30 to 230 cSt
  • Cup Number2
  • DescriptionZahn Viscosity Cup, number 2, 30 to 230 cSt, 22 to 80 sec