Bullard EVART3 EVA PAPR System with RT3 Hood; 1 Set
Representative image only.
Bullard EVART3 EVA PAPR System with RT3 Hood; 1 Set
Bullard | Mfr: EVART3 | Item: 1635917
EVA (Evolutionary Air) represents the latest in cutting-edge Powered Air-Purifying Respirator (PAPR) equipped with an innovative Active Flow Technology
₹306,000.00INR / Set of 1
₹306,000.00INR / Set of 1
Buy It Together
Current Item
Bullard EVART3 EVA PAPR System with RT3 Hood; 1 Set
Bullard EVART3 EVA PAPR System with RT3 Hood; 1 Set
1635917
₹306,000.00INR / Set of 1
+ =
Total Price 362100.00 ADD 2 ITEMS TO CART
Product Description
Includes
PAPR Assemblies include blower (EVA1), belt (EVABELT1), battery (EVABAT1), breathing tube, charger (EVASMC), airflow indicator (PA1AFI), and double-bib hood; filter is not included.
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Provides superior protection with a 1,000 APF (Assigned Protection Factor) NIOSH approved Active flow technology for Constant airflow; unique feedback loop measures flow directly with a mass flow sensor Equipped with an easy-to-read battery gauge accessible while wearing the unit Ergonomic, lightweight design makes this powered air respirator evenly balanced to provide maximum comfort Two distinct audible alarms that notify wearers of low battery and low airflow. The sound from the alarms travels through the breathing tube directly to the user’s ear A single combination filter for quick and easy installation Long-lasting 10,000-hour brushless motor that is designed for long-term reliable use Two-speed blower lets the user choose high or low speed, dependent on personal preference Filter locking tab that notifies the user that the filter is in place with both a visual indication and audible click Padded, ergonomic belt made of closed-cell, chemically resistant foam shaped to contour to the wearer’s back for extra comfort
  • 1 year warranty
More About this Item
EVA (Evolutionary Air) represents the latest in cutting-edge Powered Air-Purifying Respirator (PAPR) technology designed to provide constant air flow in a compact, streamlined form for added customer comfort.
Equipped with an innovative Active Flow Technology system that automatically maintains constant air flow, EVA is always responding to a worker’s need for more or less air.
This intelligent system is continuously working to maintain constant airflow, regardless of filter type, hood type, filter loading or battery capacity. Plus, EVA is equipped with an easy-to-read fuel gauge that lets the user quickly check the unit’s battery status.
Applications:
• Healthcare
• Pharmaceutical Manufacturing
• Paint Respirator and Coating Respirator
• Welding Respirator and Grinding Respirator
• Laboratory
• Animal Science
PAPR System, Repirator
1 Year WarrantyConformite Europeenne
Specifications
  • ComplianceNIOSH
  • DescriptionBullard® EVART3 EVA PAPR System with RT3 Hood; 1 Set