Bullard EVAHMXIS2 Hardshell Respirator with EVA System; 1 Set
Representative image only.
Bullard EVAHMXIS2 Hardshell Respirator with EVA System; 1 Set
Bullard | Mfr: EVAHMXIS2| Item: 1635919
The HMXi features all-in-one protection for your head, face, and respiratory system
₹387,000.00INR / Set of 1
₹387,000.00INR / Set of 1
Buy It Together
Current Item
Bullard EVAHMXIS2 Hardshell Respirator with EVA System; 1 Set
Bullard EVAHMXIS2 Hardshell Respirator with EVA System; 1 Set
1635919
₹387,000.00INR / Set of 1
+ =
Total Price 443100.00 ADD 2 ITEMS TO CART
Product Description
Includes
HMXi hardshell respirator (HMX-I), DuPont™ Tychem® 2000 shroud (HMXS2000), breathing tube (PAHBT), blower (EVA1), belt (EVABELT1), battery (EVABAT1), charger (EVASMC), airflow indicator (PA1AFI), disposable lens covers (HMXLC), and clear breathing tube cover (POLYBTC)
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Offers maximum head, face, and respiratory protection meeting ANSI/ISEA standard Z89.1 Type 1, Class E&G for Protective Headwear and ANSI/ISEA Z87.1 High Impact Eye and Face Protection Select models meet 1,000 Assigned Protection Factor (APF) One size fits all – no fit testing required The airflow in the HMXi is designed to rush over and around the top of the head to deliver maximum cooling to the face while also providing superior respiratory protection Best used in combination with EVA powered air-purifying respirator system Optimal Vision: Hard-coated polycarbonate lens and visors are designed to withstand the harshest environments
  • 1 year warranty
More About this Item
Applications:
• Paint Respirator
• Coating Respirator
• Welding Respirator
• Grinding Respirator
• Chemical Respirator
1 Year Warranty
Specifications
  • ComplianceNIOSH
  • DescriptionBullard® EVAHMXIS2 Hardshell Respirator with EVA System; 1 Set