Bullard HMX-I Hardshell Respirator Hood for HMXi Respirator; 1/Ea
Representative image only.
Bullard HMX-I Hardshell Respirator Hood for HMXi Respirator; 1/Ea
Bullard | Mfr: HMX-I| Item: 1635925
Hardshell respirator hood for use with HMXi Multipurpose Respirator
₹91,100.00INR / Each
₹91,100.00INR / Each
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • 1 year warranty
More About this Item
Key Features:
• HMXi offers maximum head, face, and respiratory protection meeting ANSI/ISEA standar Z89.1 Type 1, Class E&G for Protective Headwear and ANSI/ISEA Z87.1 High Impact Eye and Face Protection.
• One size fits all. No fit testing required
• The airflow in the HMXi is designed to rush over and around the top of the head to deliver maximum cooling to the face while also providing superior respiratory protection
• Panoramic Viewing Lens
• Disposable lens covers for Coatings and Painting applications
• Hard coated polycarbonate lens and visors are designed to withstand the harshest environments
2. HMXS2000 DuPont™ Tychem® Full Shroud:
Shroud for use with HMXi Hard shell respirator
Key Features:
• Full bib shroud attach within seconds to the HMXi Helmet offering coaters and painters two materials to choose from.
• DuPont™ Tychem® 2000 Chemical Resistant Full Shroud
• 1,000 Assigned Protection Factor (APF)
1 Year Warranty
Specifications
  • DescriptionBullard® HMX-I Hardshell Respirator Hood for HMXi Respirator; 1/Ea