Analtech Uniplates, 20 x 20 cm, Hard, Silica HL, 250 um Thick, 10 um Particle; 25/Pk
Representative image only.
Analtech Uniplates, 20 x 20 cm, Hard, Silica HL, 250 um Thick, 10 um Particle; 25/Pk
Analtech | Mfr: P46011| Item: 3410140
Efficient separations — economically!
₹58,400.00INR / Pkg of 25
₹58,400.00INR / Pkg of 25
Buy It Together
Current Item
Analtech Uniplates, 20 x 20 cm, Hard, Silica HL, 250 um Thick, 10 um Particle; 25/Pk
Analtech Uniplates, 20 x 20 cm, Hard, Silica HL, 250 um Thick, 10 um Particle; 25/Pk
3410140
₹58,400.00INR / Pkg of 25
+ + =
Total Price 410400.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of glass backing
More About this Item
These TLC plates combine top-quality adsorbents, consistent manufacturing, rigid QC, and careful packaging--the results of 40 years of experience dedicated to thin layer techniques. Select the perfect analytical plates for your application. Choose plates with or without an acid stable fluorescent indicator (UV 254 nm). Scored plates snap apart evenly to facilitate single-use separations or screening.
Reversed Phase plates use a readily removed nonpolar adsorbent— hydrocarbon-impregnated silica.
Specifications
  • MediaOrganic
  • Backing MaterialGlass
  • Fluorescent Indicator (° C)No
  • Particle size (micron)10
  • Plate Length (cm)20
  • Layer Thickness (micron)250
  • Plate Width (cm)20
  • DescriptionUniplates, 20x20cm/Hard/Silica HL/250um Thick/10um Particle; 25/Pk