Analtech Scraper Blades, 13 mm, Pack of five
Representative image only.
Analtech Scraper Blades, 13 mm, Pack of five
Analtech | Mfr: A05-01| Item: 3410537
Rapid removal of adsorbent from TLC plates
₹6,200.00INR / Pkg of 5
₹6,200.00INR / Pkg of 5
Product Description
Key Features
  • Optimize performance of your TLC plates
  • Constructed of durable steel
More About this Item
Scraping adsorbent layers is greatly improved with this steel flat-blade scraper. Blade is 13 mm wide and tapered to a sharp edge for rapid removal of adsorbent from plate. The scraper is available as model 34105-35 and the replacement blades for that scraper are model 34105-37.
Adsorbent scraper sold separately.
Specifications
  • DescriptionScraper Blades, 13 mm, Pack of five