Analtech TLC Adsorbent Scraper & Replacement Blades
Rapid removal of adsorbent from TLC plates
  • Optimize performance of your TLC plates
Scraping adsorbent layers is greatly improved with this steel flat-blade scraper. Blade is 13 mm wide and tapered to a sharp edge for rapid removal of adsorbent from plate. The scraper is available as model 34105-35 and the replacement blades for that scraper are model 34105-37.
6,200.00 - ₹10,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Analtech Adsorbent Scraper; 13 mm Flat-Blade 3410535 Mfr # A05-00
COMPARE
Adsorbent Scraper; 13 mm Flat-Blade
₹10,500.00INR / Each
Analtech Scraper Blades, 13 mm, Pack of five 3410537 Mfr # A05-01
COMPARE
Scraper Blades, 13 mm, Pack of five
₹6,200.00INR / Pkg of 5