Analtech Adsorbent Scraper; 13 mm Flat-Blade
Representative image only.
Analtech Adsorbent Scraper; 13 mm Flat-Blade
Analtech | Mfr: A05-00| Item: 3410535
Rapid removal of adsorbent from TLC plates
₹10,500.00INR / Each
₹10,500.00INR / Each
Product Description
Key Features
  • Optimize performance of your TLC plates
  • Constructed with durable aluminum handle
  • Extend life of the scraper with replacement blades
More About this Item
Scraping adsorbent layers is greatly improved with this steel flat-blade scraper. Blade is 13 mm wide and tapered to a sharp edge for rapid removal of adsorbent from plate. The scraper is available as model 34105-35 and the replacement blades for that scraper are model 34105-37.
Specifications
  • DescriptionAdsorbent Scraper; 13 mm Flat-Blade