Zefon High-Volume Rotary Vane Sampling Pump
Perfect pump for use with asbestos PCM and TEM cassettes, Air-O-Cell® IAQ sampling cassettes, and all viable impactors
  • Flow range of 0 to 30 L/min
  • Heavy-duty, 1/10-hp motor handles most sampling applications
89,000.00 - ₹221,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Motor hp Description
Availability Pricing
COMPARE
120 1/10 Rotary Vane Pump, High Volume; 120 VAC
₹89,000.00INR / Each
COMPARE
120 1/10 Rotary Vane Pump, with IAQ Attachments; 120 VAC
₹114,000.00INR / Each
COMPARE
120 1/10 Rotary Vane Pump, 2-Pump Kit; 120 VAC
₹221,000.00INR / Each