Zefon Filter Membrane Supports

Reinforce filter membranes to avoid membrane damage
  • Support membranes from collapsing while allowing air flow through
Cellulose pads are made up of 100% pure cellulose. Support ring meets NIOSH 5506/5515/5044 methods.
2,000.00 - ₹5,800.00INR / Pkg of 100

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Pad, Cellulose Support, 25 mm; 100/PK
₹2,000.00INR / Pkg of 100
COMPARE
Pad, Cellulose Support, 37 mm; 100/PK
₹2,100.00INR / Pkg of 100
COMPARE
Ring, Cellulose Support, 37 mm; 100/PK
₹5,800.00INR / Pkg of 100
COMPARE
Pad, Absorbent Cellulose Support, 46.5 mm; 100/PK
₹4,800.00INR / Pkg of 100
COMPARE
Pad, Cellulose Support, 47 mm; 100/PK
₹3,100.00INR / Pkg of 100
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat