Wika NSF-Certified Pressure Gauges
Manufactured with “lead free” components as defined in the SDWA
  • Made in the USA
  • Special lead-free, Silver-solder for all wetted parts
These NSF certified gauges are manufactured in accordance with NSF/ANSI 61, for lead-free content, and come in a wide range of models, sizes and configurations. The guages have special dials with NSF logo on the dial to indicate compliance. These NSF gauges are available in custom versions and configurations. Call our Application Specialists to find out about custom versions of the gauge.
4,000.00 - ₹6,900.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dial Size (in) Max Pressure (PSI) Process Connection
Availability Pricing
Wika 111.10DW NSF-Certified Pressure Gauge, 0 to 30 PSI, Bottom Mount 1/4" NPT(M); 2" Dial 6804901 Mfr # 50992563
COMPARE
2 30 1/4" NPT(M), lower
₹5,000.00INR / Each
Wika 4309546 NSF-Certified Pressure Gauge, 0 to 60 PSI, Bottom Mount 1/4" NPT(M) Connection 6804903 Mfr # 4309546
COMPARE
2 60 1/4" NPT(M), lower
₹5,100.00INR / Each
Wika 50072391 NSF-Certified Pressure Gauge, 0 to 100 PSI, Bottom Mount 1/4" NPT(M) Connection 6804905 Mfr # 50072391
COMPARE
2 100 1/4" NPT(M), lower
₹5,000.00INR / Each
Wika 4377716 NSF-Certified Pressure Gauge, 0 to 160 PSI, Bottom Mount 1/4" NPT(M); 2" Dial 6804907 Mfr # 4377716
COMPARE
2 160 1/4" NPT(M), lower
₹5,000.00INR / Each
Wika 50996993 NSF-Certified Pressure Gauge, 0 to 30 PSI, Bottom Mount 1/4" NPT(M); 4" Dial 6804909 Mfr # 50996993
COMPARE
4 30 1/4" NPT(M), lower
₹5,700.00INR / Each
Wika 50997000 NSF-Certified Pressure Gauge, 0 to 160 PSI, Bottom Mount 1/4" NPT(M); 4" Dial 6804911 Mfr # 50997000
COMPARE
4 160 1/4" NPT(M), lower
₹5,700.00INR / Each
Wika 111.12DW 2 2" NSF Dry Pressure Gauge, 0-100 psi, 1/4" Back 6804921 Mfr # 50094637
COMPARE
2 100 1/4" NPT(M), back
₹4,900.00INR / Each
Wika 4350681 2" NSF Dry Pressure Gauge, 0-160 psi, 1/4" Back 6804923 Mfr # 52578178
COMPARE
2 160 1/4" NPT(M), back
₹4,000.00INR / Each
Wika 213.53DW 2 NSF-Certified Pressure Gauge, 0 to 30 PSI, 1/4" NPT(M) Connection, Glycerin Fill 6804925 Mfr # 50135838
COMPARE
2.5 30 1/4" NPT(M), lower
₹6,600.00INR / Each
Wika 213.53DW 2 NSF-Certified Pressure Gauge, 0 to 60 PSI, 1/4" NPT(M) Connection, Glycerin Fill 6804927 Mfr # 50048813
COMPARE
2.5 60 1/4" NPT(M), lower
₹6,600.00INR / Each
Wika 213.53DW 2 2.5" NSF Filled Pressure Gauge, 0-100 psi, 1/4" Lower 6804929 Mfr # 50048805
COMPARE
2.5 100 1/4" NPT(M), lower
₹6,600.00INR / Each
Wika 213.53DW 2 2.5" NSF Filled Pressure Gauge, 0-160 psi, 1/4" Lower 6804931 Mfr # 50035428
COMPARE
2.5 160 1/4" NPT(M), lower
₹6,600.00INR / Each
Wika 213.53DW 2 2.5" NSF Filled Pressure Gauge, 0-60 psi, 1/4" Back 6804933 Mfr # 52572684
COMPARE
2.5 60 1/4" NPT(M), back
₹6,900.00INR / Each
Showing 1- 13 of 13