WHEATON Imhoff Settling Cone and Rack
Use settling cones and rack to determine settleable materials in water and wastewater samples
  • Cones are externally graduated in mL from the bottom upward
Cones (06340-02) are made of clear styrene-acrylonitrile cone features a leakproof plug for use with a heavy-gauge steel support rack (06340-20). Rack holds up to three cones and measures 17-1/2"L x 5-1/2"W x 16"H. Order racks and cones separately.
36,600.00 - ₹63,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Material
Availability Pricing
DWK Life Sciences (Kimble) Imhoff Settling Cone, Styrene-acrylonitrile, Each 0634002 Mfr # W990800
COMPARE
Imhoff Settling Cone, Styrene-acrylonitrile Styrene Acrylonitrile
₹63,600.00INR / Each
DWK Life Sciences (Kimble) 990760 Support Rack for Imhoff Settling Cones, Steel, Holds 3 Cones 0634020 Mfr # W990760
COMPARE
Support Rack for Imhoff Settling Cones, steel, holds 3 cones Steel
₹36,600.00INR / Each