DWK Life Sciences (Kimble) Imhoff Settling Cone, Styrene-acrylonitrile, Each
Representative image only.
DWK Life Sciences (Kimble) Imhoff Settling Cone, Styrene-acrylonitrile, Each
DWK Life Sciences (Kimble) | Mfr: W990800| Item: 0634002
Use settling cones and rack to determine settleable materials in water and wastewater samples
₹63,600.00INR / Each
₹63,600.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Kimble) Imhoff Settling Cone, Styrene-acrylonitrile, Each
DWK Life Sciences (Kimble) Imhoff Settling Cone, Styrene-acrylonitrile, Each
0634002
₹63,600.00INR / Each
+ =
Total Price 100200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Cones are externally graduated in mL from the bottom upward
More About this Item
Cones (06340-02) are made of clear styrene-acrylonitrile cone features a leakproof plug for use with a heavy-gauge steel support rack (06340-20). Rack holds up to three cones and measures 17-1/2"L x 5-1/2"W x 16"H. Order racks and cones separately.
Specifications
  • MaterialStyrene Acrylonitrile
  • Sampling PumpPeristaltic
  • DescriptionImhoff Settling Cone, Styrene-acrylonitrile