DWK Life Sciences (Kimble) 990760 Support Rack for Imhoff Settling Cones, Steel, Holds 3 Cones
Representative image only.
DWK Life Sciences (Kimble) 990760 Support Rack for Imhoff Settling Cones, Steel, Holds 3 Cones
DWK Life Sciences (Kimble) | Mfr: W990760| Item: 0634020
Use settling cones and rack to determine settleable materials in water and wastewater samples
₹36,600.00INR / Each
₹36,600.00INR / Each
Buy It Together
Current Item
DWK Life Sciences (Kimble) 990760 Support Rack for Imhoff Settling Cones, steel, holds 3 cones
DWK Life Sciences (Kimble) 990760 Support Rack for Imhoff Settling Cones, steel, holds 3 cones
0634020
₹36,600.00INR / Each
+ =
Total Price 100200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Cones are externally graduated in mL from the bottom upward
  • 3 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Cones (06340-02) are made of clear styrene-acrylonitrile cone features a leakproof plug for use with a heavy-gauge steel support rack (06340-20). Rack holds up to three cones and measures 17-1/2"L x 5-1/2"W x 16"H. Order racks and cones separately.
Specifications
  • MaterialSteel
  • DescriptionSupport Rack for Imhoff Settling Cones, steel, holds 3 cones
  • Warranty3 Month Product / 12 Month Parts and Labor