WHEATON® E-C Sample Glass Vials
Shrink-wrapped packaging reduces contamination
  • Constructed of either clear or amber glass
Economical screw-thread sample vials are made with either clear 33 low-extratable borosilicate glass. Choose amber colored vials for your light-sensitive materials. Vials are available with a rubber-lined phenolic cap, a PTFE-faced/rubber-lined phenolic cap, or no cap. Shrink-wrapped packaging reduces contaminates. Caps are packaged separately. Autoclavable.
17,400.00 - ₹73,700.00INR / Case of 200
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Color Tube Material Volume (mL)
Availability Pricing
DWK Life Sciences (Wheaton) 224721 E-C Sample Vials, Clear Glass with Rubber-Lined Cap, 2 mL; 200/Cs 0892310 Mfr # 224721
COMPARE
Clear Borosilicate glass 2
₹26,300.00INR / Case of 200
DWK Life Sciences (Wheaton) 224722 E-C Sample Vials, Clear Glass with Rubber-Lined Cap, 4 mL; 200/Cs 0892311 Mfr # 224722
COMPARE
Clear Borosilicate glass 4
₹26,700.00INR / Case of 200
DWK Life Sciences (Wheaton) 224724 E-C Sample Vials, Clear Glass with Rubber-Lined Cap, 8 mL; 200/Cs 0892312 Mfr # 224724
COMPARE
Clear Borosilicate glass 8
₹27,300.00INR / Case of 200
DWK Life Sciences (Wheaton) 224725 E-C Sample Vials, Clear Glass with Rubber-Lined Cap, 12 mL; 200/Cs 0892313 Mfr # 224725
COMPARE
Clear Borosilicate glass 12
₹27,300.00INR / Case of 200
DWK Life Sciences (Wheaton) 224726 E-C Sample Vials, Clear Glass with Rubber-Lined Cap, 16 mL; 200/Cs 0892314 Mfr # 224726
COMPARE
Clear Borosilicate glass 16
₹33,600.00INR / Case of 200
DWK Life Sciences (Wheaton) 224728 E-C Sample Vials, Clear Glass with Rubber-Lined Cap, 24 mL; 200/Cs 0892315 Mfr # 224728
COMPARE
Clear Borosilicate glass 24
₹41,800.00INR / Case of 200
DWK Life Sciences (Wheaton) 224731 E-C Sample Vials, Amber Glass with Rubber-Lined Cap, 2 mL; 200/Cs 0892316 Mfr # 224731
COMPARE
Clear Borosilicate glass 2
₹28,500.00INR / Case of 200
DWK Life Sciences (Wheaton) 224732 E-C Sample Vials, Amber Glass with Rubber-Lined Cap, 4 mL; 200/Cs 0892317 Mfr # 224732
COMPARE
Amber Borosilicate glass 4
₹28,800.00INR / Case of 200
DWK Life Sciences (Wheaton) 224734 E-C Sample Vials, Amber Glass with Rubber-Lined Cap, 8 mL; 200/Cs 0892318 Mfr # 224734
COMPARE
Amber Borosilicate glass 8
₹30,600.00INR / Case of 200
DWK Life Sciences (Wheaton) 224741 E-C Sample Vials, Clear Glass with PTFE/Rubber-Lined Cap, 2 mL; 200/Cs 0892320 Mfr # 224741
COMPARE
Clear Borosilicate glass 2
₹41,400.00INR / Case of 200
DWK Life Sciences (Wheaton) 224742 E-C Sample Vials, Clear Glass with  PTFE/Rubber-Lined Cap, 4 mL; 200/Cs 0892321 Mfr # 224742
COMPARE
Clear Borosilicate glass 4
₹44,900.00INR / Case of 200
DWK Life Sciences (Wheaton) 224744 E-C Sample Vials, Clear Glass with  PTFE/Rubber-Lined Cap, 8 mL; 200/Cs 0892322 Mfr # 224744
COMPARE
Clear Borosilicate glass 8
₹53,200.00INR / Case of 200
DWK Life Sciences (Wheaton) 224746 E-C Sample Vials, Clear Glass with PTFE/Rubber-Lined Cap, 16 mL; 200/Cs 0892324 Mfr # 224746
COMPARE
Clear Borosilicate glass 16
₹66,200.00INR / Case of 200
DWK Life Sciences (Wheaton) 224748 E-C Sample Vials, Clear Glass with PTFE/Rubber-Lined Cap, 24 mL; 200/Cs 0892325 Mfr # 224748
COMPARE
Clear Borosilicate glass 24
₹73,700.00INR / Case of 1
DWK Life Sciences (Wheaton) 224751 E-C Sample Vials, Amber Glass with  PTFE/Rubber-Lined Cap, 2 mL; 200/Cs 0892326 Mfr # 224751
COMPARE
Amber Borosilicate glass 2
₹52,300.00INR / Case of 200
DWK Life Sciences (Wheaton) 224752 E-C Sample Vials, Amber Glass with  PTFE/Rubber-Lined Cap, 4 mL; 200/Cs 0892327 Mfr # 224752
COMPARE
Amber Borosilicate glass 4
₹55,900.00INR / Case of 200
DWK Life Sciences (Wheaton) 224754 E-C Sample Vials, Amber Glass with  PTFE/Rubber-Lined Cap, 8 mL; 200/Cs 0892328 Mfr # 224754
COMPARE
Amber Borosilicate glass 8
₹61,200.00INR / Case of 200
DWK Life Sciences (Wheaton) 224702 E-C Sample Vials, Clear Glass without Cap, 4 mL; 200/Cs 0892331 Mfr # 224702
COMPARE
Clear Borosilicate glass 4
₹17,400.00INR / Case of 200
DWK Life Sciences (Wheaton) 224704 E-C Sample Vials, Clear Glass without Cap, 8 mL; 200/Cs 0892332 Mfr # 224704
COMPARE
Clear Borosilicate glass 8
₹20,000.00INR / Case of 200
Showing 1- 19 of 19