Vibrasorb Terrazzo Top with Vibration Damping Feet
Reduce vibration disturbances caused by nearby equipment
The black-and-white, dense terrazzo slabs feature a scratch- and chemical-resistant polished surface; supported by four vibration absorbers with neoprene feet.
389,000.00 - ₹434,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Height (in) Length (in) Width (in)
Availability Pricing
Bel-Art H18386-0000 Terrazzo Top with Vibration Damping Feet, Large 18 x 22 x 3" 0101930 Mfr # H18386-0000
COMPARE
3 18 22
₹434,000.00INR / Each
Bel-Art H18386-0001 Terrazzo Top with Vibration Damping Feet, Small 13.75 x 13.75 x 3" 0101935 Mfr # H18386-0001
COMPARE
3 13 3/4 13 3/4
₹389,000.00INR / Each