Bel-Art H18386-0000 Terrazzo Top with Vibration Damping Feet, Large 18 x 22 x 3"

Representative image only.

Bel-Art H18386-0000 Terrazzo Top with Vibration Damping Feet, Large 18 x 22 x 3"

Bel-Art | Mfr: H18386-0000| Item: 0101930

Reduce vibration disturbances caused by nearby equipment

₹434,000.00INR / Each

₹434,000.00INR / Each
Product Description

More About this Item

The black-and-white, dense terrazzo slabs feature a scratch- and chemical-resistant polished surface; supported by four vibration absorbers with neoprene feet.

Specifications
  • Height (in)3
  • Width (in)22
  • Length (in)18
  • DescriptionTerrazzo Top with Vibration Damping Feet, Large 18 x 22 x 3"
Reviews
Q&A