Bel-Art H18386-0001 Terrazzo Top with Vibration Damping Feet, Small 13.75 x 13.75 x 3"

Representative image only.

Bel-Art H18386-0001 Terrazzo Top with Vibration Damping Feet, Small 13.75 x 13.75 x 3"

Bel-Art | Mfr: H18386-0001| Item: 0101935

Reduce vibration disturbances caused by nearby equipment

₹389,000.00INR / Each

₹389,000.00INR / Each
Product Description

More About this Item

The black-and-white, dense terrazzo slabs feature a scratch- and chemical-resistant polished surface; supported by four vibration absorbers with neoprene feet.

Specifications
  • ColorBlack and White
  • Height (in)3
  • Width (in)13 3/4
  • Length (in)13 3/4
  • DescriptionTerrazzo Top with Vibration Damping Feet, Small 13.75 x 13.75 x 3"
Reviews
Q&A