Troemner Class-F Metric Heavy Capacity Cast Iron Grip Handle Weights
Acceptable for use verifying scales that have a ±0.1% accuracy or lower
  • Color coding – gold for metric
Cast Iron Grip Handle Weights larger than 5 kg/10 lb are legal for trade, which means their design is acceptable for use in checking weighing devices where the price of the item being purchased is based on its weight.
Cast Iron Grip Handle Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians. Cast Iron Grip Handle weights may be used to test most accuracy Class III scales, all scales of Class IIIL or IIII, and scales not marked with a class designation. Class F is also acceptable for use in verifying scales that have a 0.1% accuracy and lower.
19,500.00 - ₹72,100.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Class Mass Size (kg)
Availability Pricing
Troemner 9286T 25kg Class F Cast Iron Grip Handle Weight with Traceable Certificate 0102290 Mfr # 9286T
COMPARE
NIST Class F 25
₹72,100.00INR / Each
Troemner 9278-7 2kg, Class F Cast Iron Grip-Handle Weight 0102291 Mfr # 9278-7
COMPARE
None Class F 2
₹19,500.00INR / Each
Troemner 9282 10kg, Class F Cast Iron Grip-Handle Weight 0102572 Mfr # 9282
COMPARE
None Class F 10
₹37,500.00INR / Each
Troemner 9284 20kg, Cast Iron Class F Grip-Handle Calibration Mass 0102574 Mfr # 9284
COMPARE
None Class F 20
₹38,900.00INR / Each
Troemner 9286 25kg, Cast Iron Class F Grip-Handle Calibration Mass 0102576 Mfr # 9286
COMPARE
None Class F 25
₹52,300.00INR / Each
Troemner 9278-7W 2kg Class F Cast Iron Grip Handle Weight with Traceable Certificate 0102989 Mfr # 9278-7W
COMPARE
NIST Class F 2
₹31,100.00INR / Each
Troemner 9284T 20kg Cast Iron Class F Grip-Handle Calibration Mass, Traceable Certificate 1196655 Mfr # 9284T
COMPARE
NIST Class F 20
₹57,900.00INR / Each
Troemner 9282T 10kg Cast Iron Class F Grip-Handle Calibration Mass, Traceable Certificate 1196656 Mfr # 9282T
COMPARE
NIST Class F 10
₹44,800.00INR / Each
Showing 1- 8 of 8