Troemner 9282T 10kg Cast Iron Class F Grip-Handle Calibration Mass, Traceable Certificate
Representative image only.
Troemner Mfr # 9282T – Item # WW-11966-56
Acceptable for use verifying scalse that have a ±0.1% accuracy or lower
  • Color coding – gold for metric
  • 3 month warranty
PRODUCT OPTIONS
₹37,200.00INR / Each
₹13,500.00 - ₹20,400.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Mass ClassClass F
  • Mass Size (kg)10
  • Mass CertificationNIST
  • Description10kg Class F Cast Iron Grip-Handle Calibration Mass, Traceable Certificate
  • Warranty3 month
3 Month Warranty
Includes
Calibration certificate.
Key Features
  • Color coding – gold for metric
  • 3 month warranty
More About this Item
Cast Iron Grip Handle Weights larger than 5 kg/10 lb are legal for trade, which means their design is acceptable for use in checking weighing devices where the price of the item being purchased is based on its weight.
Cast Iron Grip Handle Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians. Cast Iron Grip Handle weights may be used to test most accuracy Class III scales, all scales of Class IIIL or IIII, and scales not marked with a class designation. Class F is also acceptable for use in verifying scales that have a 0.1% accuracy and lower.