Troemner Class 7 Economical Flat Balance Weights
Quality weights at an economical price
  • Adjusted to tolerances specified by ANSI/ASTM E617 Class 7
  • One-piece construction
  • One side turned up for easy handling with forceps
These flat balance weights offer quality and accuracy at an economical price. These one-piece construction flat weights are available from 1 mg to 500 mg and have one side turned up to make them easy to handle with forceps. All weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7. Weights 100 mg and below are made of aluminum; others are made of stainless steel.
Suggested markets for economical flat balance weights include, but are not limited to, educational markets.
4,100.00 - ₹9,900.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (mg)
Availability Pricing
Troemner 61010S 1 mg, Class 7 Economical Aluminum Flat Weight 0102824 Mfr # 61010S
COMPARE
None 1
₹4,100.00INR / Each
Troemner 61020S 2 mg, Class 7 Economical Aluminum Flat Weight 0102826 Mfr # 61020S
COMPARE
None 2
₹4,100.00INR / Each
Troemner 61050S 5 mg, Class 7 Economical Aluminum Flat Weight 0102828 Mfr # 61050S
COMPARE
None 5
₹4,100.00INR / Each
Troemner 61021S 20 mg, Class 7 Economical Aluminum Flat Weight 0102832 Mfr # 61021S
COMPARE
None 20
₹4,100.00INR / Each
Troemner 61051S 50 mg, Class 7 Economical Aluminum Flat Weight 0102834 Mfr # 61051S
COMPARE
None 50
₹4,100.00INR / Each
Troemner 61012S 100 mg, Class 7 Economical Aluminum Flat Weight 0102836 Mfr # 61012S
COMPARE
None 100
₹4,100.00INR / Each
Troemner 61022S 200 mg, Class 7 Economical S/S Flat Weight 0102838 Mfr # 61022S
COMPARE
None 200
₹4,100.00INR / Each
Troemner 61052S 500 mg, Class 7 Economical S/S Flat Weight 0102840 Mfr # 61052S
COMPARE
None 500
₹4,100.00INR / Each
Troemner 61012ST 100 mg, Class 7 Economical Aluminum Flat Weight with Traceable Certificate 0102872 Mfr # 61012ST
COMPARE
NIST 100
₹9,900.00INR / Each
Troemner 61052ST 500 mg, Class 7 Economical S/S Flat Weight with Traceable Certificate 0102874 Mfr # 61052ST
COMPARE
NIST 500
₹9,900.00INR / Each
Showing 1- 10 of 10