Trajan Jumbo Gas-Tight Syringes
For handling large volumes of gas
  • Heavy duty acrylic barrel
  • Easy access to sample for the addition standards or removal of subsample
  • Removeable plunger stem
7/16" UNEF thread and designed to be used with Luer Lock fittings and needles
88,100.00 - ₹137,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 009910 Jumbo Gast tight Syringe with Luer Lock, 500 mL; 1/Ea 1600206 Mfr # 009910
COMPARE
₹88,100.00INR / Each
Trajan SGE 009920 Jumbo Gast tight Syringe with Luer Lock, 1 L; 1/Ea 1600207 Mfr # 009920
COMPARE
₹92,700.00INR / Each
Trajan SGE 009930 Jumbo Gast tight Syringe with Luer Lock, 2 L; 1/Ea 1600208 Mfr # 009930
COMPARE
₹137,000.00INR / Each