Trajan SGE 009930 Jumbo Gast tight Syringe with Luer Lock, 2 L; 1/Ea
Representative image only.
Trajan SGE 009930 Jumbo Gast tight Syringe with Luer Lock, 2 L; 1/Ea
Trajan | Mfr: 009930| Item: 1600208
For handling large volumes of gas
₹137,000.00INR / Each
₹137,000.00INR / Each
Product Description
Key Features
  • Heavy duty acrylic barrel
  • Easy access to sample for the addition standards or removal of subsample
  • Removeable plunger stem
  • 6 month warranty
More About this Item
7/16" UNEF thread and designed to be used with Luer Lock fittings and needles
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Jumbo Gast tight Syringe with Luer Lock, 2 L; 1/Ea