Trajan SGE 009910 Jumbo Gast tight Syringe with Luer Lock, 500 mL; 1/Ea
Representative image only.
Trajan SGE 009910 Jumbo Gast tight Syringe with Luer Lock, 500 mL; 1/Ea
Trajan | Mfr: 009910| Item: 1600206
For handling large volumes of gas
₹88,100.00INR / Each
₹88,100.00INR / Each
Product Description
Key Features
  • Heavy duty acrylic barrel
  • Easy access to sample for the addition standards or removal of subsample
  • Removeable plunger stem
  • 6 month warranty
More About this Item
7/16" UNEF thread and designed to be used with Luer Lock fittings and needles
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Jumbo Gast tight Syringe with Luer Lock, 500 mL; 1/Ea