Trajan Fixed Cone-Tip Needle Syringes
Designed and tested to meet critical autosampler specifications
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
4,600.00 - ₹137,000.00INR / Each
41 variations of this product are available.
See All Product Options Below
41 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 001800 Fixed Needle Syringe, Compatible with Agilent Autosampler, 0.5 uL, 42 mm L, 26 G; 1/Ea 1599907 Mfr # 001800
COMPARE
₹9,900.00INR / Each
Trajan SGE 001804 Fixed Needle Syringe, Compatible with Agilent Autosampler, 0.5 uL, 42 mm L, 26 G; 6/Pk 1599908 Mfr # 001804
COMPARE
₹56,000.00INR / Pkg of 6
Trajan SGE 001810 Fixed Needle Syringe, Compatible with Agilent Autosampler, 0.5 uL, 42 mm L, 23 G; 1/Ea 1599909 Mfr # 001810
COMPARE
₹9,900.00INR / Each
Trajan SGE 001814 Fixed Needle Syringe, Compatible with Agilent Autosampler, 0.5 uL, 42 mm L, 23 G; 6/Pk 1599910 Mfr # 001814
COMPARE
₹56,000.00INR / Pkg of 6
Trajan SGE 001821 Fixed Needle Syringe, Compatible with Agilent Autosampler, 0.5uL, 42mm L, 23-26 G; 1/Ea 1599911 Mfr # 001821
COMPARE
₹9,000.00INR / Each
Trajan SGE 001822 Fixed Needle Syringe, Compatible with Agilent Autosampler, 5 uL, 42 mm L, 23-26 G; 6/Pk 1599912 Mfr # 001822
COMPARE
₹51,900.00INR / Pkg of 6
Trajan SGE 001861 Fixed Needle Syringe, Compatible with Thermo RSH Autosampler, 5 uL, 57 mm L, 23 G; 1/Ea 1599913 Mfr # 001861
COMPARE
₹12,800.00INR / Each
Trajan SGE 001865 Fixed  Needle Syringe, Compatible with Thermo RSH Autosampler, 5 uL, 57 mm L, 26 G; 1/Ea 1599915 Mfr # 001865
COMPARE
₹12,800.00INR / Each
Trajan SGE 001953 Fixed Needle Syringe, Compatible with Perkin Elmer Autosampler, 5 uL, 26 G; 1/Ea 1599921 Mfr # 001953
COMPARE
₹12,100.00INR / Each
Trajan SGE 001954 Fixed Needle Syringe, Compatible with Perkin Elmer Autosampler, 5 uL, 23 G; 1/Ea 1599922 Mfr # 001954
COMPARE
₹12,100.00INR / Each
Trajan SGE 001981 Fixed Needle Syringe, Compatible with Thermo Scientific TriPlus, 5 uL, 50 mm L, 23 G; 1/Ea 1599925 Mfr # 001981
COMPARE
₹12,000.00INR / Each
Trajan SGE 001982 Fixed Needle Syringe, Compatible with Thermo Scientific TriPlus, 5 uL, 50 mm L, 26 G; 1/Ea 1599927 Mfr # 001982
COMPARE
₹12,000.00INR / Each
Trajan SGE 0019821 Fixed Needle Syringe, Compatible with Thermo Scientific AS3000, AS2000, 5 uL, 26 G; 1/Ea 1599928 Mfr # 0019821
COMPARE
₹12,000.00INR / Each
Trajan SGE 002800 Fixed  Needle Syringe, Compatible with Agilent Autosampler, 10 uL, 42 mm L, 26 G; 1/Pk 1599963 Mfr # 002800
COMPARE
₹6,500.00INR / Each
Trajan SGE 002804 Fixed Needle Syringe, Compatible with Agilent Autosampler, 10 uL, 42 mm L, 26 G; 6/Pk 1599964 Mfr # 002804
COMPARE
₹36,100.00INR / Pkg of 6
Trajan SGE 002810 Fixed Needle Syringe, Compatible with Agilent Autosampler, 10 uL, 42 mm L, 23 G; 1/Pk 1599966 Mfr # 002810
COMPARE
₹6,400.00INR / Each
Trajan SGE 002813 Fixed Needle Syringe, Compatible with Agilent Autosampler, 10 uL, 42 mm L, 23 G; 25/Pk 1599969 Mfr # 002813
COMPARE
₹137,000.00INR / Pkg of 25
Trajan SGE 002814 Fixed Needle Syringe, Compatible with Agilent Autosampler, 10 uL, 42 mm L, 23 G; 6/Pk 1599970 Mfr # 002814
COMPARE
₹36,100.00INR / Pkg of 6
Trajan SGE 002821 Fixed Needle Syringe, Compatible with Agilent Autosampler, 10 uL, 23-26 G; 1/Ea 1599973 Mfr # 002821
COMPARE
₹6,400.00INR / Each
Trajan SGE 002822 Fixed Needle Syringe, Compatible with Agilent Autosampler, 10 uL, 23-26 G; 6/Pk 1599974 Mfr # 002822
COMPARE
₹36,100.00INR / Pkg of 6
Showing 1 - 20 of 41