Trajan SGE 001822 Fixed Needle Syringe, Compatible with Agilent Autosampler, 5 uL, 42 mm L, 23-26 G; 6/Pk
Representative image only.
Trajan SGE 001822 Fixed Needle Syringe, Compatible with Agilent Autosampler, 5 uL, 42 mm L, 23-26 G; 6/Pk
Trajan | Mfr: 001822| Item: 1599912
Designed and tested to meet critical autosampler specifications
₹51,900.00INR / Pkg of 6
₹51,900.00INR / Pkg of 6
Product Description
Key Features
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
  • 6 month warranty
More About this Item
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • Syringe TypeSGE GC Autosampler
  • Syringe Capacity (µL)5
  • Needle IncludedYes
  • Needle Tip StyleCone
  • Needle Gauge23-26
  • Needle Length (in)1.65
  • DescriptionTrajan SGE Fixed CT Needle Syringe, Compatible with Agilent Autosampler, 5uL, 42mm L, 23-26 G; 6/Pk