Trajan SGE 001810 Fixed Needle Syringe, Compatible with Agilent Autosampler, 0.5 uL, 42 mm L, 23 G; 1/Ea
Representative image only.
Trajan SGE 001810 Fixed Needle Syringe, Compatible with Agilent Autosampler, 0.5 uL, 42 mm L, 23 G; 1/Ea
Trajan | Mfr: 001810| Item: 1599909
Designed and tested to meet critical autosampler specifications
₹9,900.00INR / Each
₹9,900.00INR / Each
Product Description
Key Features
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
  • 6 month warranty
More About this Item
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Fixed CT Needle Syringe, Compatible with Agilent Autosampler, 0.5 uL, 42 mm L, 23 G; 1/Ea