Trajan Diamond Headspace Syringes
Designed and tested to meet critical autosampler specifications
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
26,000.00 - ₹28,500.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 008131 Diamond Headspace Syringe, Compatible with CTC Analytics Systems, 1mL, 23 G, SH Tip; 1/Ea 1600160 Mfr # 008131
COMPARE
₹26,000.00INR / Each
Trajan SGE 008636 Diamond Headspace Syringe, Compatible with CTC Analytics Systems, 2.5 mL, 26 G; 1/Ea 1600180 Mfr # 008636
COMPARE
₹26,000.00INR / Each
Trajan SGE 008655 Headspace GT Syringe, Compatible with Thermo RSH Autosampler,Fixed SH Needle, 2.5 mL; 1/Ea 1600181 Mfr # 008655
COMPARE
₹28,500.00INR / Each
Trajan SGE 008631 Diamond Headspace Syringe, Compatible with CTC Analytics Systems, 2.5 mL, 23 G; 1/Ea 1601154 Mfr # 008631
COMPARE
₹26,000.00INR / Each