Trajan SGE 008636 Diamond Headspace Syringe, Compatible with CTC Analytics Systems, 2.5 mL, 26 G; 1/Ea
Representative image only.
Trajan SGE 008636 Diamond Headspace Syringe, Compatible with CTC Analytics Systems, 2.5 mL, 26 G; 1/Ea
Trajan | Mfr: 008636| Item: 1600180
Designed and tested to meet critical autosampler specifications
₹26,000.00INR / Each
₹26,000.00INR / Each
Product Description
Key Features
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
  • 6 month warranty
More About this Item
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Diamond Headspace Syringe, Compatible with CTC Analytics Systems, 2.5 mL, 26 G; 1/Ea