Trajan SGE 008655 Headspace GT Syringe, Compatible with Thermo RSH Autosampler,Fixed SH Needle, 2.5 mL; 1/Ea
Representative image only.
Trajan SGE 008655 Headspace GT Syringe, Compatible with Thermo RSH Autosampler,Fixed SH Needle, 2.5 mL; 1/Ea
Trajan | Mfr: 008655| Item: 1600181
Designed and tested to meet critical autosampler specifications
₹28,500.00INR / Each
₹28,500.00INR / Each
Product Description
Key Features
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
  • 6 month warranty
More About this Item
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Headspace GT Syringe, Compatible with Thermo RSH Autosampler,Fixed SH Needle, 2.5 mL; 1/Ea