ThermoSafe® Insulated Containers
Easily store and transport dry ice, wet ice, and laboratory samples
  • Durable FDA approved high-impact double-wall polyethylene construction offer excellent durability
  • Urethane foam insulation helps maintain constant temperature inside the container
Durable insulated containers feature double-wall polyethylene shell filled with urethane foam insulation to provide outstanding temperature maintenance. Specially formulated insulation offers low sublimation rates. One-piece seamless construction ensures a tight seal and no leakage.
151,000.00 - ₹182,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Width (in)
Availability Pricing
ThermoSafe Durable Insulated Container with Drain, 2 cu ft 0372120 Mfr # HR02P-D
COMPARE
2 cu ft 19 1/4 21 1/2
₹160,000.00INR / Each
ThermoSafe Durable Insulated Container, 4 cu ft 0372121 Mfr # HR04P
COMPARE
4 cu ft 33 16
₹151,000.00INR / Each
ThermoSafe Durable Insulated Container with Casters, 4 cu ft 0372122 Mfr # HR04P-C
COMPARE
4 cu ft 33 16
₹182,000.00INR / Each