Thermo Scientific Water Analysis Accessories

Optimize water analysis
  • Choose from a variety of accessories
Get the most out of your water analysis equipment with these quality solutions and accessories.
21,200.00 - ₹81,300.00INR / Each

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific 9147BN pH electrode, 3-in-1 Gel Epoxy (Input connector: BNC) 5882292 Mfr # 9147BN
COMPARE
pH electrode, 3-in-1 Gel Epoxy (Input connector: BNC)
₹59,900.00INR / Each
Thermo Scientific 1110001 Electrode Stand For Star Series Benchtop Meters 5882350 Mfr # 090043
COMPARE
Electrode Stand For Star Series Benchtop Meters
₹81,300.00INR / Each
Thermo Scientific CODH00 Orion Mid-Range COD Kit, 0 to 1500 ppm 9958142 Mfr # CODH00
COMPARE
Orion Mid-Range COD Kit, 0 to 1500 ppm
Call For Price
Thermo Scientific CODHP0 High range COD kit, 0 to 15,000 ppm 9958144 Mfr # CODHP0
COMPARE
High range COD kit, 0 to 15,000 ppm
Call For Price
Thermo Scientific CODS01 Orion 1000 ppm COD standard, non-hazardous, 475 mL. 9958180 Mfr # CODS01
COMPARE
Orion 1000 ppm COD standard, non-hazardous, 475 mL.
₹25,400.00INR / Each
Thermo Scientific CODS10 Orion 10,000 ppm COD standard, non-hazardous, 475 mL. 9958185 Mfr # CODS10
COMPARE
Orion 10,000 ppm COD standard, non-hazardous, 475 mL.
₹21,200.00INR / Each
Thermo Scientific AC45FZ Formazin Standard Solution, 4000 NTU; 500 mL 9990032 Mfr # AC45FZ
COMPARE
Formazin Standard Solution, 4000 NTU; 500 mL
₹22,730.00INR / Each
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat