Thermo Scientific CODH00 Orion Mid-Range COD Kit, 0 to 1500 ppm
Representative image only.
Thermo Scientific CODH00 Orion Mid-Range COD Kit, 0 to 1500 ppm
Thermo Scientific | Mfr: CODH00| Item: 9958142
Optimize water analysis
Call For Price
Call For Price
Product Description
Key Features
  • Choose from a variety of accessories
More About this Item
Get the most out of your water analysis equipment with these quality solutions and accessories.
Use for EPA reporting of COD test results and general monitoring. The pre-measured reagents offer a long shelf life. Each kit contains 25 vials.
EPA United States Environmental protection agency
Specifications
  • Compatible WithThermo Scientific Orion COD Reactor & Colorimeter
  • DescriptionOrion Mid-Range COD Kit, 0 to 1500 ppm