Thermo Scientific 1110001 Electrode Stand For Star Series Benchtop Meters
Representative image only.
Thermo Scientific 1110001 Electrode Stand For Star Series Benchtop Meters
Thermo Scientific | Mfr: 090043| Item: 5882350
Make taking readings simple
₹81,300.00INR / Each
₹81,300.00INR / Each
Product Description
Key Features
  • Cable management system minimizes breakage
  • Offers 360° of motion
  • Holds up to five electrodes with 12 mm diameter
  • Aluminum construction
More About this Item
Protect and improve the use of your pH/Ion meter.
Easily move arm vertically, horizontally and front-to-back positioning into beakers or sample containers. Head holds three electrodes vertically while two additional slots hold electrodes at a 10° angle. Weighted base feature cushioning to protect table surface.
Specifications
  • Compatible WithAny benchtop meter
  • TypeStand-alone swing-arm electrode stand
  • DescriptionElectrode Stand For Star Series Benchtop Meters