Thermo Scientific™ UNIGUARD Direct Connection Guard Cartridge Holder
Provides easy column changes and eliminates the need for additional fittings
  • Easy to attach - requires no additional tubing
  • Compatible with all Thermo Scientific columns
This convenient, reusable guard cartridge holder attaches directly to the analytical column inlet and requires no additional tubing. The 1/16" male outlet fits all Thermo Scientific columns, and many additional brands as well. The 1/16" female inlet connects with a standard 1/16" nut and ferrule.
21,500.00 - ₹92,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific Uniguard LID LIFT POCKET F/33850-00 0190522 Mfr # 850-00
COMPARE
Guard Cartridge Holder - 4.0mm / 4.6mm ID
₹87,500.00INR / Each
Thermo Scientific Uniguard 850-RT Guard Cartridge Holder Standard Replacement Tip and Ferrule 0190523 Mfr # 850-RT
COMPARE
Guard Cartridge Holder Standard Replacement Tip and Ferrule
₹21,500.00INR / Each
Thermo Scientific Uniguard 852-00 Guard Cartridge Holder - 2.1mm / 3.0mm ID 0190526 Mfr # 852-00
COMPARE
Guard Cartridge Holder - 2.1mm / 3.0mm ID
₹92,900.00INR / Each