Thermo Scientific Uniguard LID LIFT POCKET F/33850-00
Representative image only.
Thermo Scientific Uniguard LID LIFT POCKET F/33850-00
Thermo Scientific | Mfr: 850-00| Item: 0190522
Provides easy column changes and eliminates the need for additional fittings
₹87,500.00INR / Each
₹87,500.00INR / Each
Product Description
Key Features
  • Easy to attach - requires no additional tubing
  • Compatible with all Thermo Scientific columns
  • 1 month warranty
More About this Item
This convenient, reusable guard cartridge holder attaches directly to the analytical column inlet and requires no additional tubing. The 1/16" male outlet fits all Thermo Scientific columns, and many additional brands as well. The 1/16" female inlet connects with a standard 1/16" nut and ferrule.
1 Month Warranty
Specifications
  • Column Length (mm)4
  • Column Inside Diameter (ID) (mm)4.6
  • DescriptionGuard Cartridge Holder - 4.0mm / 4.6mm ID
  • Warranty1 month