Thermo Scientific Uniguard 850-RT Guard Cartridge Holder Standard Replacement Tip and Ferrule
Representative image only.
Thermo Scientific Uniguard 850-RT Guard Cartridge Holder Standard Replacement Tip and Ferrule
Thermo Scientific | Mfr: 850-RT| Item: 0190523
Provides easy column changes and eliminates the need for additional fittings
₹21,500.00INR / Each
₹21,500.00INR / Each
Product Description
Key Features
  • Easy to attach - requires no additional tubing
  • Compatible with all Thermo Scientific columns
  • 1 month warranty
More About this Item
This convenient, reusable guard cartridge holder attaches directly to the analytical column inlet and requires no additional tubing. The 1/16" male outlet fits all Thermo Scientific columns, and many additional brands as well. The 1/16" female inlet connects with a standard 1/16" nut and ferrule.
1 Month Warranty
Specifications
  • DescriptionGuard Cartridge Holder Standard Replacement Tip and Ferrule
  • Warranty1 month