Thermo Scientific Orion™ Refillable Double Junction ROSS pH Electrodes
Drift less than 0.002 pH per day
  • Platinum wire prevents contamination with silver and mercury ions
  • Excellent for use in tris buffer
These provide fast, accurate, and stable pH readings. Ideal for food, pharmaceuticals, or biological samples. Double ceramic junction design protects the inner reference solution and prolongs the life of the electrode. Readings stabilize to 0.01 pH in less than 30 seconds—even with a 50degC temperature change. Electrodes read pH from 0 to 14. Max temperature is 90degC for glass electrodes and 80degC for epoxy. All electrodes have BNC connectors.
66,300.00 - ₹255,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Applications Electrode Body Material Reference Junction
Availability Pricing
Thermo Scientific 8102BN pH Probe, Refillable/DJ/Glass; BNC 0571802 Mfr # 8102BN
COMPARE
General purpose-Tris buffers Glass Ceramic
₹69,470.00INR / Each
Thermo Scientific 815600 pH Probe, Refillable/DJ/Epoxy; BNC 0571807 Mfr # 815600
COMPARE
Rugged use-TRIS buffers Epoxy Glass fiber
₹66,300.00INR / Each
Thermo Scientific 8103BN pH Probe, Refillable/DJ/Glass/Semi-Micro; BNC 0571816 Mfr # 8103BN
COMPARE
Small Samples Glass Ceramic
₹86,110.00INR / Each
Thermo Scientific 8135BN pH Probe, Refillable/DJ/Glass/Surface; BNC 0571832 Mfr # 8135BN
COMPARE
Soft, Moist surfaces, Agar plates Epoxy Glass fiber
₹214,000.00INR / Each
Thermo Scientific 8163BNWP pH Probe, Refillable/DJ/Glass/Spear-Tip; BNC 0571837 Mfr # 8163BNWP
COMPARE
General purpose-Tris buffers Glass Ceramic
₹255,000.00INR / Each
Thermo Scientific 8172BNWP Sure-Flow pH Probe, Refillable/DJ/Glass; WPBNC 0571851 Mfr # 8172BNWP
COMPARE
Dirty Samples Glass Sure-flow
₹96,340.00INR / Each
Thermo Scientific 8165BNWP Sure-Flow pH Probe, Refillable/DJ/Epoxy 0571853 Mfr # 8165BNWP
COMPARE
Dirty Samples Epoxy Sure-flow
₹95,540.00INR / Each
Thermo Scientific 8175BNWP Sure-Flow pH Probe, Refillable/DJ/Epoxy, semi-micro 0571904 Mfr # 8175BNWP
COMPARE
Dirty Samples Epoxy Sure-flow
₹131,670.00INR / Each
Showing 1- 8 of 8