Thermo Scientific 8103BN pH Probe, Refillable/DJ/Glass/Semi-Micro; BNC
Representative image only.
Thermo Scientific 8103BN pH Probe, Refillable/DJ/Glass/Semi-Micro; BNC
Thermo Scientific | Mfr: 8103BN| Item: 0571816
Drift less than 0.002 pH per day
₹86,110.00INR / Each
₹86,110.00INR / Each
Buy It Together
Current Item
Thermo Scientific 8103BN pH Probe, Refillable/DJ/Glass/Semi-Micro; BNC
Thermo Scientific 8103BN pH Probe, Refillable/DJ/Glass/Semi-Micro; BNC
0571816
₹86,110.00INR / Each
+
Thermo Scientific 810007 electrode fill solution, 3 M KCl 5/cs
0573052
₹24,600.00INR / Case of 5
=
Total Price 110710.00 ADD 2 ITEMS TO CART
Product Description
Includes
50-mL of 3 M KCl filling solution and 1-m cable. Order additional fill solution separately.
Key Features
  • Platinum wire prevents contamination with silver and mercury ions
  • Excellent for use in tris buffer
  • 1 year warranty
More About this Item
These provide fast, accurate, and stable pH readings. Ideal for food, pharmaceuticals, or biological samples. Double ceramic junction design protects the inner reference solution and prolongs the life of the electrode. Readings stabilize to 0.01 pH in less than 30 seconds—even with a 50degC temperature change. Electrodes read pH from 0 to 14. Max temperature is 90degC for glass electrodes and 80degC for epoxy. All electrodes have BNC connectors.
Models 05718-20 and 05721-26 have a pin-tip connector, which is identical to the pin-tip portion of a U.S. Standard connector.
1 Year Warranty
Specifications
  • ApplicationsSmall Samples
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference ElectrolyteROSS
  • Min pH (pH)0
  • Max pH (pH)14
  • Accuracy±0.01 pH
  • ConnectorBNC
  • Diameter (mm)6
  • Length (mm)165
  • Cable Length (m)1
  • DescriptionpH Probe, Refillable/DJ/Glass/Semi-Micro; BNC