Thermo Scientific 8175BNWP Sure-Flow pH Probe, Refillable/DJ/Epoxy, semi-micro
Representative image only.
Thermo Scientific 8175BNWP Sure-Flow pH Probe, Refillable/DJ/Epoxy, semi-micro
Thermo Scientific | Mfr: 8175BNWP| Item: 0571904
Drift less than 0.002 pH per day
₹131,670.00INR / Each
₹131,670.00INR / Each
Product Description
Includes
50-mL of 3 M KCl filling solution and 1-m cable. Order additional fill solution separately.
Key Features
  • Platinum wire prevents contamination with silver and mercury ions
  • Excellent for use in tris buffer
  • 1 year warranty
More About this Item
These provide fast, accurate, and stable pH readings. Ideal for food, pharmaceuticals, or biological samples. Double ceramic junction design protects the inner reference solution and prolongs the life of the electrode. Readings stabilize to 0.01 pH in less than 30 seconds—even with a 50degC temperature change. Electrodes read pH from 0 to 14. Max temperature is 90degC for glass electrodes and 80degC for epoxy. All electrodes have BNC connectors.
Models 05718-20 and 05721-26 have a pin-tip connector, which is identical to the pin-tip portion of a U.S. Standard connector.
1 Year Warranty
Specifications
  • ApplicationsDirty Samples
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionSure-flow
  • Reference ElectrolyteROSS
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • Cable Length (m)1
  • DescriptionSure-Flow pH Probe, Refillable/DJ/Epoxy, semi-micro